missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11926071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11926071 5 g 5g
11916071 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926071 Supplier Thermo Scientific Acros Supplier No. 444710050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 775351-57-0
Molecular Formula C13H15BFNO2
Molecular Weight (g/mol) 247.08
InChI Key RYJOVQGRIOURGT-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-4-fluorophenylboronic acid, pinacol ester, 3-cyano-4-fluorophenylboronic acid pinacol ester, benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyano-4-fluorobenzeneboronic acid pinacol ester, 3-cyano-4-fluorophenyl-boronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
PubChem CID 11402361
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N

Specifications

Melting Point 75.0°C to 76.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 247.08
Percent Purity 97%
Chemical Name or Material 3-Cyano-4-fluorophenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Cyano-4-fluorophenylboronic acid pinacol ester
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.