missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzeneboronic acid, 98%

Product Code. 11375425
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375425 1 g 1g
11395425 250 mg 250mg
11385425 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375425 Supplier Thermo Scientific Alfa Aesar Supplier No. L19635.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150255-96-2 | C7H6BNO2 | 146.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150255-96-2
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318967
InChI Key XDBHWPLGGBLUHH-UHFFFAOYSA-N
Synonym 3-cyanophenyl boronic acid, 3-cyanobenzeneboronic acid, 3-boronobenzonitrile, 3-dihydroxyboranyl benzonitrile, 3-cyanobenzene boronic acid, 3-cyano-phenyl-boronic acid, boronic acid, 3-cyanophenyl, pubchem1804, phenylboronic acid, 4
PubChem CID 2734325
IUPAC Name (3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)C#N

Specifications

Melting Point ∽298°C (decomposition)
Quantity 1 g
Beilstein 6136720
Formula Weight 146.94
Percent Purity ≥98%
Chemical Name or Material 3-Cyanobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.