missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzyl Bromide, TRC

Product Code. 30356338
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30356338

Brand: Toronto Research Chemicals TRCC96236525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyanobenzyl Bromide
CAS 28188-41-2
Molecular Formula C8H6BrN
Quantity 25 g
Synonym 1-Bromomethyl-3-cyanobenzene, 3-(Bromomethyl)benzonitrile, NSC 108298, m-(Bromomethyl)benzonitrile, m-Cyano-α-bromotoluene, m-Cyanobenzyl bromide, α-Bromo-m-tolunitrile
InChI Formula InChI=1S/C8H6BrN/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5H2
SMILES BrCC1=CC(C#N)=CC=C1
IUPAC Name 3-(bromomethyl)benzonitrile
Molecular Weight (g/mol) 196.04
Formula Weight 194.9684
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.