missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenyl isocyanate, 97%

Product Code. 11357426
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357426 5 g 5g
11347426 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357426 Supplier Thermo Scientific Alfa Aesar Supplier No. L09432.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16413-26-6 | C8H4N2O | 144.133 g/mol

3-Cyanophenyl isocyanate serves as a starting reagent in the synthesis of R/S-4-(3-chlorophenylaminocarbonylamino)-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran and 1-(3-cyanophenyl)-3-(2-methoxy-6-pentadecylbenzyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyanophenyl isocyanate serves as a starting reagent in the synthesis of R/S-4-(3-chlorophenylaminocarbonylamino)-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran and 1-(3-cyanophenyl)-3-(2-methoxy-6-pentadecylbenzyl)urea.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16413-26-6
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
MDL Number MFCD00013856
InChI Key NZHPVPMRNASEQK-UHFFFAOYSA-N
Synonym 3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
PubChem CID 2733315
IUPAC Name 3-isocyanatobenzonitrile
SMILES C1=CC(=CC(=C1)N=C=O)C#N

Specifications

Melting Point 51°C to 54°C
Odor Irritating
Quantity 5 g
UN Number UN2811
Beilstein 3089117
Sensitivity Moisture sensitive
Formula Weight 144.13
Percent Purity 97%
Chemical Name or Material 3-Cyanophenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.