missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)trichlorosilane, 98%

Product Code. 11499050
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499050 25 g 25g
11409060 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499050 Supplier Thermo Scientific Alfa Aesar Supplier No. A13787.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1071-27-8 | C4H6Cl3NSi | 202.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-27-8
Molecular Formula C4H6Cl3NSi
Molecular Weight (g/mol) 202.534
MDL Number MFCD00013832
InChI Key HMFFOEBLYHLRQN-UHFFFAOYSA-N
Synonym 3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane
PubChem CID 14063
IUPAC Name 4-trichlorosilylbutanenitrile
SMILES C(CC#N)C[Si](Cl)(Cl)Cl

Specifica

Boiling Point 237°C to 238°C
Quantity 25 g
Flash Point 92°C (198°F)
Odor Pungent
Refractive Index 1.463
Sensitivity Moisture sensitive
Density 1.28
Linear Formula Cl3Si(CH2)3CN
UN Number UN2987
Beilstein 906924
Formula Weight 202.54
Percent Purity 98%
Chemical Name or Material (3-Cyanopropyl)trichlorosilane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.