missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopropyldimethylchlorosilane, 95%

Product Code. 10792952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10792952 10 mL Glass bottle 10mL
10793722 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792952 Supplier Thermo Scientific Acros Supplier No. 313380100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18156-15-5 | C6H12ClNSi | 161.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18156-15-5
Molecular Formula C6H12ClNSi
Molecular Weight (g/mol) 161.7
InChI Key GPIARXZSVWTOMD-UHFFFAOYSA-N
Synonym 3-cyanopropyl dimethylchlorosilane, 4-chlorodimethylsilyl butanenitrile, 3-cyanopropyldimethylchlorosilane, chloro 3-cyanopropyl dimethylsilane, butanenitrile, 4-chlorodimethylsilyl, 4-chlorodimethylsilyl butyronitrile, 4-chloro dimethyl silyl butanenitrile, cyanopropyldimethylchlorosilane, chlorocyanopropyldimethylsilane, cyanopropyl dimethylchlorosilane
PubChem CID 87478
IUPAC Name 4-[chloro(dimethyl)silyl]butanenitrile
SMILES C[Si](C)(CCCC#N)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Colorless
Density 0.9900g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2Si(Cl)(CH2)3CN
Refractive Index 1.4450 to 1.4470 (20°C, 589nm)
Quantity 10 mL
Specific Gravity 0.99
Solubility Information Solubility in water: reacts.
Formula Weight 161.7
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Cyanopropyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Cyanopropyldimethylchlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.