missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 10157513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157513 1 g Glass bottle 1g
10033530 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157513 Supplier Thermo Scientific Acros Supplier No. 294390010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4771-80-6 | Molecular Formula: C7H10O2 | Molecular Weight: 126.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4771-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
MDL Number MFCD00013781
InChI Key VUSWCWPCANWBFG-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
PubChem CID 20903
IUPAC Name cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 17.0°C
Color Blue-Gray to Brown-Yellow
Density 1.0800g/mL
Boiling Point 238.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Quantity 1 g
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 126.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Cyclohexenecarboxylic acid, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Cyclohexenecarboxylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.