missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propyne, 97%

Code produit 11434517
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
25 g
Conditionnement:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
11434517 25 g 25g
11414517 1 g 1g
11424517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11434517 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur B20505.14
 Restricted Item
Estimated Shipment: 02-12-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 17715-00-3 | C9H14 | 122.211 g/mol

3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 17715-00-3
Molecular Formula C9H14
Molecular Weight (g/mol) 122.211
MDL Number MFCD00041562
InChI Key UARFKZSJGDQRLF-UHFFFAOYSA-N
Synonym 3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
PubChem CID 87265
IUPAC Name prop-2-ynylcyclohexane
SMILES C#CCC1CCCCC1

Spécification

Density 0.845
Boiling Point 157°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.462
Quantity 25 g
UN Number UN3295
Beilstein 1849402
Solubility Information Slightly soluble in water.
Formula Weight 122.21
Percent Purity 97%
Chemical Name or Material 3-Cyclohexyl-1-propyne
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.