missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid, 99%, BLD Pharm™

Product Code. 31424773
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31424773 100 g
31424758 1 kg
31418569 25 g
31415285 10 g
31420629 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31424773 Supplier BLD Pharm Supplier No. BD119879100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 73463-39-5
Molecular Formula C9H19NO4S
MDL Number MFCD00041778
Quantity 100 g
SMILES OC(CNC1CCCCC1)CS(O)(=O)=O
Molecular Weight (g/mol) 237.32
Percent Purity 99%
Chemical Name or Material 3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.