missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexylpropionic acid, 98+%

Product Code. 11415647
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415647 250 g 250g
11405647 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415647 Supplier Thermo Scientific Alfa Aesar Supplier No. B23613.30
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 701-97-3 | C9H16O2 | 156.23 g/mol

3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

Solubility
Soluble in water 10g/ L.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-97-3
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00001527
InChI Key HJZLEGIHUQOJBA-UHFFFAOYSA-N
Synonym cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
PubChem CID 69702
IUPAC Name 3-cyclohexylpropanoic acid
SMILES OC(=O)CCC1CCCCC1

Specifications

Melting Point 14°C to 17°C
Density 0.99
Boiling Point 276°C
Flash Point 130°C (266°F)
Odor Unpleasant
Refractive Index 1.4636
Quantity 250 g
Beilstein 2042799
Solubility Information Soluble in water 10g/ L.
Formula Weight 156.23
Percent Purity ≥98%
Chemical Name or Material 3-Cyclohexylpropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.