missing translation for 'onlineSavingsMsg'
Learn More

3-Deazaneplanocin A hydrochloride, MedChemExpress

Product Code. 16465008
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465008 10 mg 10mg
16447737 5 mg 5mg
16431229 1 mg 1mg
16413286 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465008 Supplier Medchem Express Supplier No. HY12186
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.

3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.

Specifications

Chemical Name or Material 3-Deazaneplanocin A hydrochloride
CAS 120964-45-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C12H15ClN4O3
Quantity 10 mg
Synonym DZNep hydrochloride NSC 617989 hydrochloride 3-Deazaneplanocin hydrochloride
Solubility Information H2O : 50 mg/mL (167.38 mM; Need ultrasonic)
SMILES O[C@@H]1[C@H](O)C(CO)=C[C@H]1N2C=NC3=C2C=CN=C3N.[H]Cl
Molecular Weight (g/mol) 298.73
Formula Weight 298.73
Percent Purity 98.37%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.