missing translation for 'onlineSavingsMsg'
Learn More

3-Demethyl Thiocolchicine 2-O-(2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester), TRC

Product Code. 30352828
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352828 5 mg 5mg
30350822 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352828 Supplier Toronto Research Chemicals Supplier No. TRCD2313055MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Demethyl Thiocolchicine 2-O-(2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester)
CAS 250662-47-6
Molecular Formula C34H39NO14S
Quantity 5 mg
Synonym (7S)-7-(Acetylamino)-5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-3-yl 2,3,4-Tri-O-acetyl-beta-D-glucopyranosiduronic Acid Methyl Ester
InChI Formula InChI=1S/C34H39NO14S/c1-15(36)35-22-11-9-19-13-24(27(42-5)28(43-6)26(19)20-10-12-25(50-8)23(40)14-21(20)22)48-34-32(47-18(4)39)30(46-17(3)38)29(45-16(2)37)31(49-34)33(41)44-7/h10,12-14,22,29-32,34H,9,11H2,1-8H3,(H,35,36)/t22-,29+,30+,31+,32-,34-/m1/s1
SMILES COC(=O)[C@H]1O[C@@H](Oc2cc3CC[C@@H](NC(=O)C)C4=CC(=O)C(=CC=C4c3c(OC)c2OC)SC)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC Name methyl (2S,3S,4S,5R,6S)-6-[[(7R)-7-acetamido-1,2-dimethoxy-10-methylsulfanyl-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-3-yl]oxy]-3,4,5-triacetyloxyoxane-2-carboxylate
Molecular Weight (g/mol) 717.74
Formula Weight 717.2091
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.