missing translation for 'onlineSavingsMsg'
Learn More

3'-Deoxyadenosine, 98%

Product Code. 15466859
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466859 100 mg 100mg
15446859 10 mg 10mg
15456859 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466859 Supplier Thermo Scientific Alfa Aesar Supplier No. J65366.MC
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73-03-0 | C10H13N5O3 | 251.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-03-0
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00037998
InChI Key OFEZSBMBBKLLBJ-PNFQIUPPNA-N
Synonym cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8
PubChem CID 6303
ChEBI CHEBI:29014
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 100 mg
UN Number UN2811
Beilstein 35194
Merck Index 14,2529
IUPAC Name (2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
Formula Weight 251.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.