missing translation for 'onlineSavingsMsg'
Learn More

3-Desacetyl Vecuronium Bromide, TRC

Product Code. 30358684
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358684 1 mg 1mg
30352987 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358684 Supplier Toronto Research Chemicals Supplier No. TRCD2887151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73319-13-8
Molecular Formula C32 H55 N2 O3 . Br
Molecular Weight (g/mol) 595.6947
Synonym Piperidinium, 1-[(2β,3α,5α,16β,17β)-17-(acetyloxy)-3-hydroxy-2-(1-piperidinyl)androstan-16-yl]-1-methyl-, bromide (1:1), Piperidinium, 1-[(2β,3α,5α,16β,17β)-17-(acetyloxy)-3-hydroxy-2-(1-piperidinyl)androstan-16-yl]-1-methyl-, bromide (9CI), 3-Hydroxyvecuronium bromide, Org 7268, 3-Desacetyl Vecuronium Bromide, Vecuronium Bromide Related Compound F (USP)
IUPAC Name [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-hydroxy-10,13-dimethyl-16-(1-methylpiperidin-1-ium-1-yl)-2-piperidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;bromide
SMILES [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCCCC5)[N+]6(C)CCCCC6

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C32H55N2O3.BrH/c1-22(35)37-30-28(34(4)17-9-6-10-18-34)20-26-24-12-11-23-19-29(36)27(33-15-7-5-8-16-33)21-32(23,3)25(24)13-14-31(26,30)2;/h23-30,36H,5-21H2,1-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,27-,28-,29-,30-,31-,32-;/m0./s1
Formula Weight 594.3396
Chemical Name or Material 3-Desacetylvecuronium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.