missing translation for 'onlineSavingsMsg'
Learn More

3-Desmethyl Sulfentrazone, TRC

Product Code. 30133660
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133660 5 mg 5mg
30136988 10 mg 10mg
30012041 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133660 Supplier Toronto Research Chemicals Supplier No. TRCD2957205MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134391-02-9
Molecular Formula C10 H8 Cl2 F2 N4 O3 S
Molecular Weight (g/mol) 373.16
Synonym Methanesulfonamide, N-[2,4-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]-, N-[2,4-Dichloro-5-[4-(difluoromethyl)-4,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]methanesulfonamide
IUPAC Name N-[2,4-dichloro-5-[4-(difluoromethyl)-5-oxo-1,2,4-triazol-1-yl]phenyl]methanesulfonamide
SMILES CS(=O)(=O)Nc1cc(N2N=CN(C(F)F)C2=O)c(Cl)cc1Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H8Cl2F2N4O3S/c1-22(20,21)16-7-3-8(6(12)2-5(7)11)18-10(19)17(4-15-18)9(13)14/h2-4,9,16H,1H3
Formula Weight 371.9662
Chemical Name or Material Sulfentrazone-desmethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.