missing translation for 'onlineSavingsMsg'
Learn More

3-(Di-tert-butylphosphonium)propane sulfonate, 97%

Product Code. 15497187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497187 1 g 1g
15438855 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497187 Supplier Thermo Scientific Alfa Aesar Supplier No. H34253.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1055888-89-5 | C11H25O3PS | 268.352 g/mol

3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1055888-89-5
Molecular Formula C11H25O3PS
Molecular Weight (g/mol) 268.352
MDL Number MFCD12546062
InChI Key JPNPRWMRUCIEMN-UHFFFAOYSA-N
Synonym 3-di-t-butylphosphonium propane sulfonate, 3-di-tert-butylphosphino propane-1-sulfonic acid, dtbpps, di-t-butyl 3-sulfonatopropyl phosphine, 3-di-tert-butylphosphonium propane sulfonate, 3-di-tert-butylphosphanyl propane-1-sulfonic acid
PubChem CID 24949188
IUPAC Name 3-ditert-butylphosphanylpropane-1-sulfonic acid
SMILES CC(C)(C)P(CCCS(=O)(=O)O)C(C)(C)C

Specifications

Linear Formula [(CH3)3C]2P(CH2)3SO3H
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 268.35
Percent Purity 97%
Chemical Name or Material 3-(Di-tert-butylphosphonium)propane sulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.