missing translation for 'onlineSavingsMsg'
Learn More

3-Diethoxyphosphoryloxy-1,2,3-benzotriazin-4(3H)-one, 98%

Product Code. 11485840
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485840 25 g 25g
11475840 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485840 Supplier Thermo Scientific Alfa Aesar Supplier No. H26481.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 165534-43-0 | C11H14N3O5P | 299.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165534-43-0
Molecular Formula C11H14N3O5P
Molecular Weight (g/mol) 299.223
MDL Number MFCD01236967
InChI Key AJDPNPAGZMZOMN-UHFFFAOYSA-N
Synonym depbt, 3-diethoxyphosphoryloxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryloxy-1,2,3-benzotrizin-4 3h-one, diethyl 4-oxobenzo d 1,2,3 triazin-3 4h-yl phosphate, diethyl 4-oxo-1,2,3-benzotriazin-3-yl phosphate, 3-diethoxyphosphoryloxy-3h-benzo d 1,2,3 triazin-4-one, 3-diethoxyphosphinyl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxy-phosphoryloxy-3h-benzo d 1,2,3 triazin-4-one
PubChem CID 4293995
IUPAC Name diethyl (4-oxo-1,2,3-benzotriazin-3-yl) phosphate
SMILES CCOP(=O)(OCC)ON1C(=O)C2=CC=CC=C2N=N1

Specifications

Melting Point 71°C to 76°C
Quantity 25 g
Formula Weight 299.22
Percent Purity 98%
Chemical Name or Material 3-Diethoxyphosphoryloxy-1,2,3-benzotriazin-4(3H)-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.