missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one, 98%

Product Code. 11976721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976721 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11976721 Supplier Thermo Scientific Acros Supplier No. 441530050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 165534-43-0 | C11H14N3O5P | 299.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 165534-43-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 73.0°C to 77.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C11H14N3O5P
Quantity 5 g
Synonym depbt, 3-diethoxyphosphoryloxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryloxy-1,2,3-benzotrizin-4 3h-one, diethyl 4-oxobenzo d 1,2,3 triazin-3 4h-yl phosphate, diethyl 4-oxo-1,2,3-benzotriazin-3-yl phosphate, 3-diethoxyphosphoryloxy-3h-benzo d 1,2,3 triazin-4-one, 3-diethoxyphosphinyl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxy-phosphoryloxy-3h-benzo d 1,2,3 triazin-4-one
InChI Key AJDPNPAGZMZOMN-UHFFFAOYSA-N
SMILES CCOP(=O)(OCC)ON1C(=O)C2=CC=CC=C2N=N1
IUPAC Name diethyl (4-oxo-1,2,3-benzotriazin-3-yl) phosphate
Molecular Weight (g/mol) 299.23
PubChem CID 4293995
Formula Weight 299.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(Diethoxyphosphoryloxy)-1, 2, 3-benzotriazin-4(3H)-one
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.