missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propyl isothiocyanate, 97%

Product Code. 11362976
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362976 5 g 5g
11352976 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362976 Supplier Thermo Scientific Alfa Aesar Supplier No. L10750.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2626-52-0 | C8H16N2S | 172.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2626-52-0
Molecular Formula C8H16N2S
Molecular Weight (g/mol) 172.29
MDL Number MFCD00041133
InChI Key QPBVKSLJBRCKIF-UHFFFAOYSA-N
Synonym 3-diethylamino propyl isothiocyanate, diethyl 3-isothiocyanatopropyl amine, 3-diethylamino propylisothiocyanate, diethyl-3-isothiocyanato-propyl-amine, acmc-20aniz, n,n-diethyl-3-isothiocyanato-propan-1-amine, 3-diethylaminopropyl isothiocyanate, 3-diethylamino propanisothiocyanate, 3-diethylamino-1-propyl isothiocyanate, 3-n,n-diethylamino propylisothiocyanate
PubChem CID 2758240
IUPAC Name N,N-diethyl-3-isothiocyanatopropan-1-amine
SMILES CCN(CC)CCCN=C=S

Specifications

Density 0.95
Boiling Point 94°C to 95°C (3.5 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.503
Quantity 5 g
UN Number UN2922
Beilstein 1757446
Sensitivity Moisture sensitive
Formula Weight 172.29
Percent Purity 97%
Chemical Name or Material 3-Diethylamino-1-propyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.