missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethylamino)-7-[2-[4-[[4-[(2,5-dioxo-1-pyrrolidinyl)oxy]-4-oxobutyl]methylamino]phenyl]diazenyl]-5-phenylphenazinium Hexafluorophosphate, TRC

Product Code. 30381998
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381998 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381998 Supplier Toronto Research Chemicals Supplier No. TRCD8712405MG
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Diethylamino)-7-(2-(4-((4-((2,5-dioxo-1-pyrrolidinyl)oxy)-4-oxobutyl)methylamino)phenyl)diazenyl)-5-phenylphenazinium Hexafluorophosphate
CAS 871240-95-8
Molecular Formula C37 H38 N7 O4 . F6 P
Quantity 5 mg
Synonym 3-(Diethylamino)-7-[2-[4-[[4-[(2,5-dioxo-1-pyrrolidinyl)oxy]-4-oxobutyl]methylamino]phenyl]diazenyl]-5-phenylphenazinium Hexafluorophosphate
InChI Formula InChI=1S/C37H38N7O4.F6P/c1-4-42(5-2)30-18-20-32-34(25-30)43(29-10-7-6-8-11-29)33-24-27(15-19-31(33)38-32)40-39-26-13-16-28(17-14-26)41(3)23-9-12-37(47)48-44-35(45)21-22-36(44)46;1-7(2,3,4,5)6/h6-8,10-11,13-20,24-25H,4-5,9,12,21-23H2,1-3H3;/q+1;-1
SMILES CCN(CC)c1ccc2nc3ccc(cc3[n+](c4ccccc4)c2c1)\N=N\c5ccc(cc5)N(C)CCCC(=O)ON6C(=O)CCC6=O.F[P-](F)(F)(F)(F)F
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[4-[(E)-[8-(diethylamino)-10-phenylphenazin-10-ium-2-yl]diazenyl]-N-methylanilino]butanoate;hexafluorophosphate
Molecular Weight (g/mol) 789.706
Formula Weight 789.263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.