missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylaminophenol, 99%

Product Code. 10031540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10031540 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10031540 Supplier Thermo Scientific Acros Supplier No. 146631000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-68-9 | C10H15NO | 165.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91-68-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Brown-Gray or Red
Boiling Point 170°C (15.0 mmHg)
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H15NO
Linear Formula (C2H5)2NC6H4OH
MDL Number MFCD00002265
Quantity 100 g
Beilstein 13, 408
Synonym 3-diethylamino phenol, phenol, 3-diethylamino, n,n-diethyl-m-aminophenol, n,n-diethyl-3-aminophenol, m-diethylamino phenol, 3-hydroxy-n,n-diethylaniline, phenol, m-diethylamino, n,n-diethyl-3-hydroxyaniline, unii-bqy98q7yu5, ccris 4615
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols,ethers,ketones,esters,,carbon disulfide,caustic soda,alkalis and,aromatic hydrocarbons,insoluble in ligroin
InChI Key WAVOOWVINKGEHS-UHFFFAOYSA-N
SMILES CCN(CC)C1=CC=CC(O)=C1
IUPAC Name 3-(diethylamino)phenol
Molecular Weight (g/mol) 165.24
PubChem CID 7062
Formula Weight 165.23
Percent Purity 99%
Physical Form Flakes or Granules
Chemical Name or Material 3-Diethylaminophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Diethylaminophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.