missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylaminopropylamine, 99+%

Product Code. 10375790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10375790 250 mL Glass bottle 250mL
10466910 1 L Glass bottle 1 L
10096203 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10375790 Supplier Thermo Scientific Supplier No. 114092500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-78-9 | C7H18N2 | 130.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-78-9
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008218
InChI Key QOHMWDJIBGVPIF-UHFFFAOYSA-N
Synonym 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
PubChem CID 61011
IUPAC Name N',N'-diethylpropane-1,3-diamine
SMILES CCN(CC)CCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -60°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 168°C to 171°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2N(CH2)3NH2
Refractive Index 1.4406 to 1.4426
Quantity 250 mL
Specific Gravity 0.82
Solubility Information Solubility in water: miscible with water
Viscosity 1.3 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Diethylaminopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Diethylaminopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.