missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1-(4-methoxyphenyl)-2-propen-1-one, 95%, Thermo Scientific™

Product Code. 15498285
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498285 25 g 25g
15488285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498285 Supplier Thermo Scientific Alfa Aesar Supplier No. H33051.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18096-70-3
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.26
MDL Number MFCD00097914,MFCD00097914
InChI Key FXNAUCNJLHEGGF-UHFFFAOYSA-N
Synonym 2z-3-dimethylamino-1-4-methoxyphenyl prop-2-en-1-one, 3-dimethylamino-1-4-methoxy-phenyl-propenone
PubChem CID 17024616
IUPAC Name (Z)-3-(dimethylamino)-1-(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(C=C1)C(=O)C=CN(C)C

Specifications

Melting Point 92°C to 95°C
Quantity 25 g
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 3-Dimethylamino-1-(4-methoxyphenyl)-2-propen-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.