missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals

Product Code. 15490919
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490919 1 g 1g
15410929 250 mg 250mg
15400929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490919 Supplier Thermo Scientific Alfa Aesar Supplier No. H63939.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3414-64-0
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.244
MDL Number MFCD00544197
InChI Key SVNVKGIWGOHXPW-UHFFFAOYSA-N
Synonym 5-nitrogramine, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine, 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, indole, 3-dimethylamino methyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, dimethyl 5-nitro-1h-indol-3-yl methyl amine, 4-22-00-04317 beilstein handbook reference, 3-dimethylaminomethyl-5-nitroindole, 1h-indole-3-methanamine,n-dimethyl-5-nitro
PubChem CID 18882
IUPAC Name N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
SMILES CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Beilstein 216692
Formula Weight 219.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Dimethylaminomethyl)-5-nitroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.