missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminopropyl chloride hydrochloride, 98%

Product Code. 11492817
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492817 500 g 500g
11482817 100 g 100g
11402827 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492817 Supplier Thermo Scientific Alfa Aesar Supplier No. A15459.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5407-04-5 | C5H13Cl2N | 158.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-5-5407
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
MDL Number MFCD00012521
InChI Key LJQNMDZRCXJETK-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride
PubChem CID 94308
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCCl.Cl

Specifications

Melting Point 138°C to 144°C
Quantity 500 g
Beilstein 3670046
Sensitivity Hygroscopic
Formula Weight 158.07
Percent Purity 98%
Chemical Name or Material 3-Dimethylaminopropyl chloride hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.