missing translation for 'onlineSavingsMsg'
Learn More

3-(Diphenylphosphino)propylamine, 97+%

Product Code. 15407158
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407158 1 g 1g
15427158 250 mg 250mg
15417158 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15407158 Supplier Thermo Scientific Alfa Aesar Supplier No. H60672.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16605-03-1 | C15H18NP | 243.29 g/mol

3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16605-03-1
Molecular Formula C15H18NP
Molecular Weight (g/mol) 243.29
MDL Number MFCD00233832
InChI Key DQZWMOWSTWWMPP-UHFFFAOYSA-N
Synonym 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine
PubChem CID 280677
IUPAC Name 3-diphenylphosphanylpropan-1-amine
SMILES NCCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Linear Formula (C6H5)2PCH2CH2CH2NH2
Quantity 1 g
UN Number UN2735
Beilstein 2730850
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 243.28
Percent Purity ≥97%
Chemical Name or Material 3-(Diphenylphosphino)propylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.