missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-cyclohexen-1-one, 99%

Product Code. 10614962 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10614962

Brand: Thermo Scientific Acros 117880250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5323-87-5 | C8H12O2 | 140.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5323-87-5
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001580
InChI Key JWCFJPLIRVYENQ-UHFFFAOYSA-N
Synonym 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
PubChem CID 79216
IUPAC Name 3-ethoxycyclohex-2-en-1-one
SMILES CCOC1=CC(=O)CCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Peach to White
Density 1.0400g/mL
Boiling Point 76°C to 78°C (1.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H7(=O)
Refractive Index 1.5030 to 1.505
Quantity 25 g
Specific Gravity 1.04
Solubility Information Solubility in water: slightly soluble
Formula Weight 140.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxy-2-cyclohexen-1-one
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.