missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-hydroxybenzaldehyde, 97%

Product Code. 13488378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13488378 500 g Glass bottle 500g
10081760 5 g Glass bottle 5g
10286150 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13488378 Supplier Thermo Scientific Acros Supplier No. 156035000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-32-4 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
Synonym ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
PubChem CID 8467
ChEBI CHEBI:48408
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H3(OH)CHO
Quantity 500 g
Beilstein 08,256
Merck Index 15,3908
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,glycerol and prpylene,glycol
Formula Weight 166.18
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Ethoxy-4-hydroxybenzaldehyde, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethoxy-4-hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.