missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzaldehyde, 97%

Product Code. 11433188
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433188 10 g 10g
11423188 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433188 Supplier Thermo Scientific Alfa Aesar Supplier No. A18940.09
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22924-15-8 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22924-15-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00016606
InChI Key QZMGMXBYJZVAJN-UHFFFAOYSA-N
Synonym m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde
PubChem CID 89908
IUPAC Name 3-ethoxybenzaldehyde
SMILES CCOC1=CC=CC(=C1)C=O

Specifications

Density 1.07
Boiling Point 132°C to 134°C (15 mmHg)
Quantity 10 g
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 3-Ethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.