missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxysalicylaldehyde, 97%

Product Code. 11445546
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11445546 50 g 50g
11455546 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 11445546

Brand: Thermo Scientific Alfa Aesar B20702.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 492-88-6 | C9H10O3 | 166.176 g/mol

3-Ethoxysalicylaldehyde is employed in the preparation of Schiff base ligand through condensation with trans-1,2-diaminocyclohexane. It is also useful for the preparation of substituted salen-type Schiff base ligands. Further, it is used to prepare 2-ethoxy-6-[(3-methylaminopropylimino)methyl]phenol by reaction with N-methylpropane-1,3-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxysalicylaldehyde is employed in the preparation of Schiff base ligand through condensation with trans-1,2-diaminocyclohexane. It is also useful for the preparation of substituted salen-type Schiff base ligands. Further, it is used to prepare 2-ethoxy-6-[(3-methylaminopropylimino)methyl]phenol by reaction with N-methylpropane-1,3-diamine.

Solubility
Soluble in cold water. Insoluble in methanol, ethanol, acetone and acetonitrile

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-88-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003323
InChI Key OFQBYHLLIJGMNP-UHFFFAOYSA-N
Synonym 3-ethoxysalicylaldehyde, benzaldehyde, 3-ethoxy-2-hydroxy, novovanillin, o-ethylvanillin, salicylaldehyde, 3-ethoxy, unii-hb4h48mkni, 2-hydroxy-3-ethoxybenzaldehyde, hb4h48mkni, 3-ethoxy-2-hydroxy-benzaldehyde, pubchem3123
PubChem CID 68117
IUPAC Name 3-ethoxy-2-hydroxybenzaldehyde
SMILES CCOC1=CC=CC(=C1O)C=O

Specifications

Melting Point 66°C to 68°C
Boiling Point 263°C to 264°C
Quantity 50 g
Sensitivity Air Sensitive
Solubility Information Soluble in cold water. Insoluble in methanol,ethanol,acetone and acetonitrile
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 3-Ethoxysalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.