missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxythiophene-2-carboxylic acid, 97%

Product Code. 11479806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479806 1 g 1g
11489806 5 g 5g
11499806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479806 Supplier Thermo Scientific Alfa Aesar Supplier No. B20296.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 139926-23-1 | C7H8O3S | 172.198 g/mol

3-Ethoxythiophene-2-carboxylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxythiophene-2-carboxylic acid is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139926-23-1
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.198
MDL Number MFCD00205205
InChI Key FTKDCOINUAYGTC-UHFFFAOYSA-N
PubChem CID 2777652
IUPAC Name 3-ethoxythiophene-2-carboxylic acid
SMILES CCOC1=C(SC=C1)C(=O)O

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Synonym maybridge1_003798, acmc-20an4i, 3-ethoxy-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid,3-ethoxy, 2-thiophenecarboxylicacid, 3-ethoxy, 2-thiophenecarboxylic acid, 3-ethoxy
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material 3-Ethoxythiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.