missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-1-pentyn-3-ol, 98%

Product Code. 11382466
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382466 25 g 25g
11372466 5 g 5g
11392466 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382466 Supplier Thermo Scientific Alfa Aesar Supplier No. L06335.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6285-06-9 | C7H12O | 112.17 g/mol

3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6285-06-9
MDL Number MFCD00014392

Specifications

Density 0.873
Boiling Point 138°C to 140°C
Flash Point 38°C (100°F)
Molecular Formula C7H12O
Refractive Index 1.438
Quantity 25 g
UN Number UN1987
Beilstein 1740944
Synonym 3-ethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3-ethyl, diethylethynylcarbinol, 3-pentanol, 3-ethynyl, diaethyl-aethenyl-carbinol german, 3-aethyl-pentin-1-ol-3, 3-aethyl-pentin-1-ol-3 german, d iethylethynylcarbinol, acmc-1bagf, diaethyl-aethenyl-carbinol
Solubility Information Not miscible in water.
InChI Key PUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILES CCC(CC)(C#C)O
IUPAC Name 3-ethylpent-1-yn-3-ol
Molecular Weight (g/mol) 112.17
PubChem CID 22698
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.