missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methyl-1-pentene, 99%, Thermo Scientific Chemicals

Product Code. 11449047
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449047 5 g 5g
11459047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449047 Supplier Thermo Scientific Alfa Aesar Supplier No. B20624.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19780-66-6
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00048664
InChI Key HPHHYSWOBXEIRG-UHFFFAOYSA-N
Synonym 3-ethyl-2-methyl-1-pentene, 1-pentene, 3-ethyl-2-methyl, 2-methyl-3-ethyl-1-pentene, acmc-1bqij, 1-pentene,3-ethyl-2-methyl
PubChem CID 140591
IUPAC Name 3-ethyl-2-methylpent-1-ene
SMILES CCC(CC)C(=C)C

Specifications

Density 0.72
Refractive Index 1.4124
Quantity 5 g
UN Number UN3295
Formula Weight 112.22
Percent Purity 99%
Chemical Name or Material 3-Ethyl-2-methyl-1-pentene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.