missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methylbenzoxazolium iodide, 95%, Thermo Scientific™

Product Code. 11483058
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483058 100 g 100g
11463058 5 g 5g
11473058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483058 Supplier Thermo Scientific Alfa Aesar Supplier No. A18778.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5260-37-7
Molecular Formula C10H12INO
Molecular Weight (g/mol) 289.12
MDL Number MFCD00044141
InChI Key UZWYLWSUBQEUMC-UHFFFAOYSA-M
Synonym 3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade
PubChem CID 21331
IUPAC Name 3-ethyl-2-methyl-1,3-benzoxazol-3-ium;iodide
SMILES [I-].CC[N+]1=C(C)OC2=CC=CC=C12

Specifications

Melting Point 196°C to 198°C (decomposition)
Quantity 100 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 289.12
Percent Purity 95%
Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.