missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one ,95%, Thermo Scientific™

Product Code. 10649934
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10649934 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10649934 Supplier Thermo Scientific Maybridge Supplier No. BTB13417DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 71556-74-6
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C15H21NO2
MDL Number MFCD00276840
Quantity 1 g
Synonym 3-ethyl-3-3-hydroxyphenyl-1-methylazepan-2-one, 3-ethylhexahydro-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one, 3-ethyl-hexahydro-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one, 3-ethyl-3-hydroxyphenyl-1-methylazepan-2-one, 2h-azepin-2-one,3-ethylhexahydro-3-3-hydroxyphenyl-1-methyl, 3-ethyl-3-3-hydroxy-phenyl-1-methyl-azepan-2-one, hexahydro-3-ethyl-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one
InChI Key YFSJZMANJNLCOQ-UHFFFAOYSA-N
SMILES CCC1(CCCCN(C1=O)C)C2=CC(=CC=C2)O
IUPAC Name 3-ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Molecular Weight (g/mol) 247.338
PubChem CID 2794573
Formula Weight 247.34
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.