missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide, TRC

Product Code. 30374037
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374037 25 g
30388334 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374037 Supplier Toronto Research Chemicals Supplier No. TRCE67885025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54016-70-5
Molecular Formula C8H14BrNOS
Molecular Weight (g/mol) 252.17
SMILES [Br-].CC[n+]1csc(CCO)c1C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H14NOS.BrH/c1-3-9-6-11-8(4-5-10)7(9)2;/h6,10H,3-5H2,1-2H3;1H/q+1;/p-1
Formula Weight 250.9979
Chemical Name or Material 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.