missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethyl-d5-toluene, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Ethyltoluene-D5 (ethyl-D5) |
| CAS | 1398065-64-9 |
| Molecular Formula | C9 D5 H7 |
| Quantity | 0.25 g |
| Synonym | 3-Ethyl-d5-toluene, 3-Ethyltoluene-D5 (ethyl-D5), 3-Ethyltoluene-d5, 1-Methyl-3-(ethyl-d5)benzene |
| InChI Formula | InChI=1 S/C9H12/c1-3-9-6-4-5-8(2)7-9/h4-7 H,3H2,1-2H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])c1cccc(C)c1 |
| IUPAC Name | 1-methyl-3-(1,1,2,2,2-pentadeuterioethyl)benzene |
| Molecular Weight (g/mol) | 125.2224 |
| Formula Weight | 125.1253 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction