missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylrhodanine, 98%

Product Code. 11436236
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436236 5 g 5g
11446236 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436236 Supplier Thermo Scientific Alfa Aesar Supplier No. B23047.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7648-01-3 | C7H3Br2F3 | 303.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7648-01-3

Specifications

Melting Point 36°C to 39°C
Density 1.303
Boiling Point 127°C to 129°C (4 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C7H3Br2F3
MDL Number MFCD00013554
Quantity 5 g
Synonym 1,4-dibromo-2-trifluoromethyl benzene, 2,5-dibromobenzotrifluoride, benzene, 1,4-dibromo-2-trifluoromethyl, 2,5-dibromo trifluoromethyl benzene, 1,4-dibromo-2-trifluoromethyl-benzene, toluene, 2,5-dibromo-alpha,alpha,alpha-trifluoro, pubchem9537, 2,5-dibromobenzotrifloride, 2,5-dibromo-, ,-trifluorotoluene
InChI Key VWKFJAOCLPPQGR-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC(Br)=CC=C1Br
IUPAC Name 1,4-dibromo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 303.90
PubChem CID 82108
Formula Weight 161.25
Percent Purity 98%
Chemical Name or Material 3-Ethylrhodanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.