missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethylthio)propanol, 97%, Thermo Scientific™

Product Code. 11405367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405367 25 g 25g
11495357 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405367 Supplier Thermo Scientific Alfa Aesar Supplier No. B22710.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18721-61-4
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00236041
InChI Key KRUJXSIEMOWXOF-UHFFFAOYSA-N
Synonym 3-ethylthio propanol, 1-propanol, 3-ethylthio, 3-ethylthio propan-1-ol, 3-ethylsulfanyl propan-1-ol, 3-ethylthiopropanol, 3-ethylthiopropan-1-ol, 3-ethylthio-1-propanol, acmc-1c82g, 3-ethylsulfanyl-propan-1-ol
PubChem CID 87762
IUPAC Name 3-ethylsulfanylpropan-1-ol
SMILES CCSCCCO

Specifications

Density 0.992
Boiling Point 227°C to 228°C
Flash Point 75°C (167°F)
Refractive Index 1.483
Quantity 25 g
Beilstein 1732293
Formula Weight 120.21
Percent Purity 97%
Chemical Name or Material 3-(Ethylthio)propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.