missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylpyridine, 97%

Product Code. 15450308
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450308 25 g 25g
15430308 1 g 1g
15440308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450308 Supplier Thermo Scientific Alfa Aesar Supplier No. H53465.14
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2510-23-8 | C7H5N | 103.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2510-23-8
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD02177459
InChI Key CLRPXACRDTXENY-UHFFFAOYSA-N
Synonym pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene
PubChem CID 186003
IUPAC Name 3-ethynylpyridine
SMILES C#CC1=CN=CC=C1

Specifications

Melting Point 39°C to 40°C
Boiling Point 83°C to 84°C (30 mmHg)
Flash Point 57°C (135°F)
Quantity 25 g
UN Number UN1325
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material 3-Ethynylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.