missing translation for 'onlineSavingsMsg'
Learn More

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%

Product Code. 10571311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10571311 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571311 Supplier Thermo Scientific Acros Supplier No. 265690010
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 92511-32-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 261.0°C to 263.0°C
CAS 92511-32-5
CAS Min % 97.5
CAS Max % 100.0
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H11NO2
Packaging Glass Bottle
MDL Number MFCD00143158
Synonym diendo-3-amino-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, rac-1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-endo-aminobicyclo 2.2.1 hept-5-ene-2-endo-carboxylic acid, 1r,2r,3s,4s/1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 1r,1beta,4s,4beta-3alpha-amino-2alpha-norborna-5-enecarboxylic acid, 1r,2s,3 r,4s-3-amino-5-bicyclo 2,2,1 heptene-2-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylicacid, 3-amino-, 1r,2s,3r,4s, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol
InChI Key FCYFJGGJCJDCPB-BNHYGAARSA-N
SMILES C1C2C=CC1C(C2C(=O)O)N
IUPAC Name (1R,2S,3R,4S)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Molecular Weight (g/mol) 153.18
Quantity 1 g
PubChem CID 13160880
Formula Weight 153.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.