missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-iodopyridine, 97%, Thermo Scientific Chemicals

Product Code. 10109799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10109799 1 g 1g
10715462 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10109799 Supplier Thermo Scientific Acros Supplier No. 436380010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-75-3
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.98
InChI Key MSLUTDVOKZOXTA-UHFFFAOYSA-N
Synonym 3-fluoro-4-iodo-pyridine, 4-iodo-3-fluoropyridine, pyridine, 3-fluoro-4-iodo, pubchem9249, acmc-1ccsk, pyridine,3-fluoro-4-iodo, 3-fluoro-4-iodopyridine, 3-fluoro-4-iodopyridine 1g
PubChem CID 10998636
IUPAC Name 3-fluoro-4-iodopyridine
SMILES C1=CN=CC(=C1I)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 189.4°C to 190.4°C
Color Cream to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 222.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoro-4-iodopyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.