missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethoxy)benzaldehyde, Maybridge

Product Code. 10577590
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10577590 1 g Amber Glass Bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10577590

Brand: Thermo Scientific Maybridge MAY00275DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 473917-15-6
Packaging Amber Glass Bottle
Molecular Formula C8H4F4O2
Quantity 1 g
Synonym 3-fluoro-4-trifluoromethoxy benzaldehyde, 3-fluoro-4-trifluoromethoxy-benzaldehyde
InChI Key RQLRUBHAAVGRPW-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=O)F)OC(F)(F)F
IUPAC Name 3-fluoro-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol) 208.112
PubChem CID 2783358
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 3-fluoro-4-(trifluoromethoxy)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.