missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-5'-(pentafluorothio)acetophenone, 97%, Thermo Scientific™

Product Code. 15456117
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456117 1 g 1g
15498385 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456117 Supplier Thermo Scientific Alfa Aesar Supplier No. H33302.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1240257-57-1
Molecular Formula C8H6F6OS
Molecular Weight (g/mol) 264.185
MDL Number MFCD16652464
InChI Key XABWPYOWUIIPAB-UHFFFAOYSA-N
Synonym 3'-fluoro-5'-pentafluorosulfur acetophenone, 1-3-fluoro-5-pentafluoro-??-sulfanyl phenyl ethanone, 3'-fluoro-5'-pentafluorothio acetophenone
PubChem CID 66523548
IUPAC Name 1-[3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC(=C1)S(F)(F)(F)(F)F)F

Especificaciones

Quantity 1 g
Formula Weight 264.18
Percent Purity 97%
Chemical Name or Material 3'-Fluoro-5'-(pentafluorothio)acetophenone
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.