missing translation for 'onlineSavingsMsg'
Learn More

(3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid, 97%, BLD Pharm™

Product Code. 30776916
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30776916 25 g 25g
30774112 5 g 5g
30773313 10 g 10g
30774253 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30776916 Supplier BLD Pharm Supplier No. BD1032025g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 159020-59-4
Molecular Formula C7H5BF4O2
MDL Number MFCD06657735
Quantity 25 g
SMILES OB(O)C1=CC(F)=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 207.92
Percent Purity 97%
Chemical Name or Material (3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.