missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-trifluoromethylbenzoic acid, 98%, BLD Pharm™

Product Code. 30784644
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30784644 25 g 25g
30777419 500 g 500g
30781417 10 g 10g
30778952 100 g 100g
30784561 5 g 5g
30776814 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30784644 Supplier BLD Pharm Supplier No. BD1084525g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 161622-05-5
Molecular Formula C8H4F4O2
MDL Number MFCD00061293
Quantity 25 g
SMILES O=C(O)C1=CC(C(F)(F)F)=CC(F)=C1
Molecular Weight (g/mol) 208.11
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-trifluoromethylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.