missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals

Product Code. 11381287
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11381287

Brand: Thermo Scientific Alfa Aesar L01479.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.

Solubility
Soluble in water (28 g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-90-7
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00063075
InChI Key VIIAUOZUUGXERI-UHFFFAOYNA-N
Synonym 3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine
PubChem CID 92100
ChEBI CHEBI:32771
IUPAC Name 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(F)=C1)C(O)=O

Specifications

Melting Point ∽280°C (decomposition)
Beilstein 3204804
Solubility Information Soluble in water (28g/L).
Quantity 500 mg
Formula Weight 199.18
Percent Purity 98%
Chemical Name or Material 3-Fluoro-DL-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.