missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-methylaniline, 97%

Product Code. 11495137
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495137 10 g 10g
11405147 50 g 50g
11415147 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495137 Supplier Thermo Scientific Alfa Aesar Supplier No. B22565.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1978-37-6 | C7H8FN | 125.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978-37-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD02683095
InChI Key FHYDHJXZZQCXOX-UHFFFAOYSA-N
Synonym n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl
PubChem CID 2759011
IUPAC Name 3-fluoro-N-methylaniline
SMILES CNC1=CC(=CC=C1)F

Specifications

Density 1.11
Boiling Point 94°C (19 mmHg)
Flash Point 86°C (187°F)
Odor Characteristic
Quantity 10 g
UN Number UN2810
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-N-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.