missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoroacetophenone, 99%, Thermo Scientific™

Product Code. 10646242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646242 5 g Glass bottle 5g
10636242 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646242 Supplier Thermo Scientific Acros Supplier No. 204440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificatori chimici

CAS 455-36-7
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000338
InChI Key HCEKGPAHZCYRBZ-UHFFFAOYSA-N
Synonym 3'-fluoroacetophenone, 1-3-fluorophenyl ethanone, 3-fluoroacetophenone, m-fluoroacetophenone, ethanone, 1-3-fluorophenyl, 1-3-fluorophenyl ethan-1-one, acetophenone, 3'-fluoro, 3'-fluoro-acetophenone, 1-acetyl-3-fluorobenzene
PubChem CID 9967
SMILES CC(=O)C1=CC=CC(F)=C1

Specifica

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 81.0°C (9.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4COCH3
Refractive Index 1.5080 to 1.5100
Quantity 5 g
Beilstein 07, III, 961
IUPAC Name 1-(3-fluorophenyl)ethan-1-one
Formula Weight 138.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3′-Fluoroacetophenone
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.