missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroazetidine hydrochloride, 95%

Product Code. 15425788
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425788 500 mg 500mg
15415788 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425788 Supplier Thermo Scientific Alfa Aesar Supplier No. H59454.ME
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 617718-46-4 | C3H7ClFN | 111.544 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617718-46-4
Molecular Formula C3H7ClFN
Molecular Weight (g/mol) 111.544
MDL Number MFCD05663715
InChI Key PXFUWRWCKSLCLS-UHFFFAOYSA-N
Synonym 3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt
PubChem CID 10125054
IUPAC Name 3-fluoroazetidine;hydrochloride
SMILES C1C(CN1)F.Cl

Specifications

Quantity 500 mg
Formula Weight 111.55
Percent Purity 95%
Chemical Name or Material 3-Fluoroazetidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.